Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fibp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172334.2 | NP_758837.1 | 3 | 404 | ||
XM_006230701.2 | XP_006230763.1 | 3 | 460 | |||
GenBank mRNA | AY093421.1 | - | 3 | 405 | ||
BC061802.1 | - | 3 | 425 | 5598566 | ||
FQ228181.1 | - | 3 | 456 | |||
FQ234469.1 | - | 3 | 456 | |||
FQ234564.1 | - | 3 | 456 | |||
FQ235188.1 | - | 2 | 381 |