Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019370.2 | NP_062243.2 | 16 | 1513 | ||
GenBank mRNA | BC097326.1 | - | 16 | 1513 | 7458431 | |
D30649.1 | - | 16 | 1540 | |||
U78787.1 | - | 16 | 1510 | |||
U78788.1 | - | 16 | 1516 | |||
Z47987.1 | - | 16 | 1497 |