Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCR2 SIRNA Assays ›
Gene Symbol:
Ccr2
Gene Name:
chemokine (C-C motif) receptor 2
Gene Aliases:
-
Chromosome Location:
-
UniGene ID:
Rn.211983
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
191997
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ccr2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_021866.1
NP_068638.1
2
1070
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
blood vessel remodeling
inflammatory response to antigenic stimulus
monocyte chemotaxis
positive regulation of T-helper 1 type immune response
negative regulation of type 2 immune response
cellular calcium ion homeostasis
inflammatory response
immune response
humoral immune response
cellular defense response
G-protein coupled receptor signaling pathway
regulation of vascular endothelial growth factor production
positive regulation of T cell chemotaxis
negative regulation of angiogenesis
cytokine-mediated signaling pathway
sensory perception of pain
cellular homeostasis
hemopoiesis
regulation of cell migration
positive regulation of interferon-gamma production
positive regulation of interleukin-2 production
positive regulation of tumor necrosis factor production
T-helper 17 cell chemotaxis
positive regulation of tumor necrosis factor biosynthetic process
negative regulation of eosinophil degranulation
positive regulation of MAPK cascade
positive regulation of alpha-beta T cell proliferation
positive regulation of inflammatory response
positive regulation of T cell activation
response to hyperoxia
chemokine-mediated signaling pathway
positive regulation of monocyte chemotaxis
positive regulation of immune complex clearance by monocytes and macrophages
positive regulation of monocyte extravasation
positive regulation of CD8-positive, alpha-beta T cell extravasation
positive regulation of astrocyte chemotaxis
positive regulation of hematopoietic stem cell migration
C-C chemokine receptor activity
cytokine binding
chemokine binding
C-C chemokine binding
CCR2 chemokine receptor binding
chemokine (C-C motif) ligand 2 binding
chemokine (C-C motif) ligand 12 binding
chemokine (C-C motif) ligand 7 binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-jmz56:80/10.239.163.191:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0