Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NISCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276293.1 | NP_001263222.1 | 5, 6 | 715 | ||
NM_001276294.1 | NP_001263223.1 | 5, 6 | 715 | |||
NM_007184.3 | NP_009115.2 | 5, 6 | 715 | |||
XM_005264839.2 | XP_005264896.1 | 4, 5 | 488 | |||
GenBank mRNA | AB023192.1 | - | 6, 7 | 653 | ||
AF082516.1 | - | 5, 6 | 607 | |||
AK001505.1 | - | 3, 4 | 374 | |||
AK027428.1 | - | 5, 6 | 593 | |||
AK291398.1 | - | 5, 6 | 619 | |||
AK315001.1 | - | 5, 6 | 619 | |||
BC038102.2 | - | 5, 6 | 619 | 5268247 | ||
BC054494.1 | - | 5, 6 | 613 | 6195532 | ||
BC056900.1 | - | 5, 6 | 585 | 6143316 |