Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prelp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053385.1 | NP_445837.1 | 2 | 1022 | ||
XM_006249890.2 | XP_006249952.1 | 2 | 1276 | |||
XM_006249891.2 | XP_006249953.1 | 3 | 1620 | |||
XM_008769571.1 | XP_008767793.1 | 2 | 1290 | |||
XM_008769572.1 | XP_008767794.1 | 3 | 1635 | |||
XM_008769573.1 | XP_008767795.1 | 2 | 1377 | |||
GenBank mRNA | AF163569.1 | - | 2 | 1022 | ||
BC072487.1 | - | 2 | 1040 | 7104853 |