Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113521.1 | NP_001106993.1 | 4 | 544 | ||
NM_001113522.1 | NP_001106994.1 | 4 | 544 | |||
NM_053740.2 | NP_446192.2 | 1 | 178 | |||
XM_008771376.1 | XP_008769598.1 | 4 | 11516 | |||
XM_008771377.1 | XP_008769599.1 | 4 | 11515 | |||
XM_008771378.1 | XP_008769600.1 | 4 | 11516 | |||
XM_008771379.1 | XP_008769601.1 | 601 | ||||
XM_008771380.1 | XP_008769602.1 | 514 | ||||
GenBank mRNA | AF044673.1 | - | 1 | 178 | ||
AY034823.1 | - | 1 | 178 | |||
AY996221.1 | - | 1 | 178 | |||
BC131844.1 | - | 2 | 231 |