Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133535.2 | NP_598219.2 | 3, 4 | 355 | ||
XM_006231191.2 | XP_006231253.1 | 3, 4 | 443 | |||
XM_006231192.2 | XP_006231254.1 | 3, 4 | 443 | |||
XM_006231193.2 | XP_006231255.1 | 3, 4 | 443 | |||
XM_008760303.1 | XP_008758525.1 | 3, 4 | 443 | |||
GenBank mRNA | AF353305.1 | - | 3, 4 | 411 | ||
BC086376.1 | - | 3, 4 | 355 | 7135348 |