Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vimp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173120.2 | NP_775143.2 | 5 | 490 | ||
GenBank mRNA | AF367467.1 | - | 5 | 500 | ||
BC059111.1 | - | 5 | 494 | 6920258 | ||
FQ210095.1 | - | 5 | 482 | |||
FQ218853.1 | - | 5 | 500 | |||
FQ219944.1 | - | 5 | 482 | |||
FQ220082.1 | - | 5 | 485 | |||
FQ225575.1 | - | 5 | 500 | |||
FQ229642.1 | - | 5 | 482 |