Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022008.1 | NP_071291.1 | 4, 5 | 274 | ||
GenBank mRNA | AW914994.1 | - | 4, 5 | 274 |