Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scamp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031725.1 | NP_113913.1 | 7 | 1409 | ||
XM_006240999.1 | XP_006241061.1 | 7 | 1429 | |||
GenBank mRNA | AF285631.1 | - | 7 | 1409 | ||
BC090342.1 | - | 7 | 1404 | 7304207 |