Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp27b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053763.1 | NP_446215.1 | 2, 3 | 393 | ||
XM_008765374.1 | XP_008763596.1 | 3, 4 | 2265 | |||
GenBank mRNA | AB001992.1 | - | 2, 3 | 393 | ||
AF000139.1 | - | 2, 3 | 385 |
Gene Symbol: LOC100909462 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773774.1 | XP_008771996.1 | 1153 |