Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012134.2 | NP_036266.2 | 1 | 454 | ||
GenBank mRNA | AK126474.1 | - | 1 | 320 | ||
AK295824.1 | - | 1 | 414 | |||
AK304852.1 | - | 1 | 373 | |||
AK311740.1 | - | 1 | 382 | |||
BC001755.1 | - | 1 | 335 | |||
BC080187.1 | - | 1 | 403 | 6177902 | ||
DB228115.1 | - | 1 | 454 | |||
X54162.1 | - | 1 | 334 |