Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012895.3 | NP_037027.2 | 6 | 552 | ||
XM_006251631.1 | XP_006251693.1 | 6 | 666 | |||
GenBank mRNA | BC081712.1 | - | 6 | 552 | 7193044 | |
FQ209450.1 | - | 1 | 21 | |||
FQ218369.1 | - | 2 | 215 | |||
FQ225093.1 | - | 2 | 223 | |||
U90340.1 | - | 6 | 555 |