Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021579.1 | NP_067590.1 | 4 | 483 | ||
GenBank mRNA | AF093139.1 | - | 4 | 535 |