Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012334.2 | NP_036466.2 | 5 | 1037 | ||
XM_006714475.2 | XP_006714538.1 | 5 | 813 | |||
GenBank mRNA | AB018342.2 | - | 5 | 1024 | ||
AF132021.1 | - | 5 | 649 | |||
AF234532.1 | - | 5 | 791 | |||
AF247457.2 | - | 5 | 722 | |||
AK125246.1 | - | 5 | 1021 | |||
BC014956.1 | - | 5 | 1033 | |||
BC071965.1 | - | 4 | 592 | |||
BC094838.1 | - | 4 | 592 | |||
BC137168.1 | - | 5 | 746 | 9021165 | ||
BC150285.1 | - | 5 | 1024 | 8860110 |