Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031533.5 | NP_113721.4 | 6 | 1561 | ||
GenBank mRNA | BC089792.1 | - | 6 | 1544 | 7370057 | |
DQ351215.1 | - | 6 | 1902 | |||
X03478.1 | - | 6 | 1450 |
Gene Symbol: Ugt2b17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769997.1 | XP_008768219.1 | 6 | 1901 | ||
XM_008769998.1 | XP_008768220.1 | 6 | 2658 |