Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012093.3 | NP_036225.2 | 6 | 1746 | ||
NM_174858.2 | NP_777283.1 | 6 | 1135 | |||
XM_005270739.4 | XP_005270796.1 | 6 | 982 | |||
XM_006710572.3 | XP_006710635.1 | 6 | 913 | |||
XM_017001008.1 | XP_016856497.1 | 6 | 930 | |||
XM_017001009.1 | XP_016856498.1 | 6 | 1638 | |||
XM_017001010.1 | XP_016856499.1 | 6 | 1965 | |||
XM_017001011.1 | XP_016856500.1 | 6 | 748 | |||
XM_017001012.1 | XP_016856501.1 | 6 | 1204 | |||
GenBank mRNA | AF445193.1 | - | 6 | 1135 | ||
AY171600.2 | - | 6 | 1751 | |||
BC012467.2 | - | 1 | 98 | |||
BC033896.1 | - | 6 | 1751 | 4818069 | ||
BC036666.1 | - | 6 | 1061 | 4816351 | ||
DN993295.1 | - | 3 | 323 |