Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FADS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178128.5 | NP_835229.3 | 3 | 752 | ||
XM_005257224.4 | XP_005257281.2 | 3 | 746 | |||
XM_017024458.1 | XP_016879947.1 | 4 | 527 | |||
GenBank mRNA | AK094411.1 | - | 3 | 600 | ||
AK293720.1 | - | 1 | 299 | |||
AY203929.1 | - | 3 | 561 | |||
BC117230.1 | - | 3 | 581 | 40125781 |