Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145134.1 | NP_001138606.1 | 5, 6 | 736 | ||
NM_001145135.1 | NP_001138607.1 | 6, 7 | 838 | |||
NM_001145137.1 | NP_001138609.1 | 5, 6 | 862 | |||
NM_004377.3 | NP_004368.1 | 6, 7 | 799 | |||
NM_152245.2 | NP_689451.1 | 6, 7 | 838 | |||
NM_152246.2 | NP_689452.1 | 6, 7 | 799 | |||
GenBank mRNA | AK223174.1 | - | 6, 7 | 738 | ||
AK291316.1 | - | 6, 7 | 763 | |||
AK297866.1 | - | 5, 6 | 648 | |||
AK299395.1 | - | 6, 7 | 775 | |||
BC142954.1 | - | 5, 6 | 805 | 8860341 | ||
D87812.1 | - | 5, 6 | 719 | |||
U62733.1 | - | 5, 6 | 746 | |||
U66828.1 | - | 6, 7 | 738 | |||
Y08682.1 | - | 6, 7 | 751 | |||
Y08683.1 | - | 6, 7 | 751 |
Gene Symbol: CHKB-CPT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027928.2 | 16, 17 | 2270 | |||
GenBank mRNA | AB051457.1 | - | 13, 14 | 2908 | ||
BC131570.1 | - | 8, 9 | 1051 | 40108355 |