Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR2DS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012313.1 | NP_036445.1 | 3, 4 | 389 | ||
GenBank mRNA | AF022047.1 | - | 3, 4 | 382 | ||
AM851002.1 | - | 3, 4 | 414 | |||
AY789063.2 | - | 2, 3 | 362 | |||
L76670.1 | - | 3, 4 | 389 | |||
X97231.1 | - | 3, 4 | 437 |
Gene Symbol: KIR2DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014219.2 | NP_055034.2 | 3, 4 | 407 | ||
GenBank mRNA | AF285434.1 | - | 2, 3 | 338 | ||
AY366242.1 | - | 3, 4 | 407 | |||
KP260924.1 | - | 3, 4 | 374 | |||
L76669.1 | - | 3, 4 | 389 | |||
U24075.1 | - | 3, 4 | 392 |
Gene Symbol: KIR2DL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015868.2 | NP_056952.2 | 3, 4 | 415 | ||
GenBank mRNA | AF022048.1 | - | 3, 4 | 378 | ||
AF285435.1 | - | 2, 3 | 338 | |||
AY789057.1 | - | 3, 4 | 378 | |||
BC032422.1 | - | 3, 4 | 391 | 5220339 | ||
BC050730.1 | - | 3, 4 | 409 | 5219336 | ||
BC110613.1 | - | 3, 4 | 392 | 40031619 | ||
FN395076.1 | - | 3, 4 | 378 | |||
FN552550.1 | - | 3, 4 | 360 | |||
L41268.1 | - | 3, 4 | 415 | |||
L76662.1 | - | 3, 4 | 389 | |||
L76663.1 | - | 3, 4 | 389 | |||
U24074.1 | - | 3, 4 | 399 | |||
U73395.1 | - | 3, 4 | 399 | |||
U96190.1 | - | 1, 2 | 315 |
Gene Symbol: KIR2DL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080770.1 | NP_001074239.1 | 3, 4 | 409 | ||
NM_001080772.1 | NP_001074241.1 | 3, 4 | 409 | |||
NM_002255.5 | NP_002246.5 | 3, 4 | 409 | |||
GenBank mRNA | AF002979.1 | - | 3, 4 | 369 | ||
AF002980.1 | - | 3, 4 | 381 | |||
AF002981.1 | - | 3, 4 | 381 | |||
AF002982.1 | - | 3, 4 | 381 | |||
AF034771.1 | - | 3, 4 | 375 | |||
AF034772.1 | - | 3, 4 | 375 | |||
AF034773.1 | - | 3, 4 | 375 | |||
AF276292.1 | - | 3, 4 | 369 | |||
AF285436.1 | - | 3, 4 | 396 | |||
AK309531.1 | - | 4, 5 | 645 | |||
AY223512.2 | - | 1, 2 | 296 | |||
AY223513.1 | - | 1, 2 | 296 | |||
AY223514.1 | - | 1, 2 | 296 | |||
AY223515.1 | - | 1, 2 | 296 | |||
AY250088.1 | - | 1, 2 | 296 | |||
AY601812.1 | - | 3, 4 | 363 | |||
AY789058.1 | - | 3, 4 | 363 | |||
AY789059.1 | - | 3, 4 | 363 | |||
AY789060.1 | - | 3, 4 | 363 | |||
AY789061.1 | - | 3, 4 | 363 | |||
AY789062.1 | - | 3, 4 | 363 | |||
BC041611.1 | - | 3, 4 | 410 | 5485491 | ||
BM918119.1 | - | 409 | ||||
EU194340.1 | - | 3, 4 | 369 | |||
EU194342.1 | - | 3, 4 | 369 | |||
U71199.1 | - | 3, 4 | 381 | |||
U73394.1 | - | 3, 4 | 381 | |||
X97229.1 | - | 3, 4 | 374 | |||
X99479.1 | - | 3, 4 | 369 | |||
X99480.1 | - | 3, 4 | 380 | |||
Y13054.1 | - | 3, 4 | 369 |
Gene Symbol: KIR2DS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291695.1 | NP_001278624.1 | 3, 4 | 413 | ||
NM_001291701.1 | NP_001278630.1 | 3, 4 | 413 | |||
NM_012312.4 | NP_036444.1 | 3, 4 | 413 | |||
GenBank mRNA | AF285438.1 | - | 2, 3 | 338 | ||
AJ002102.1 | - | 3, 4 | 378 | |||
AJ002103.1 | - | 3, 4 | 378 | |||
AJ002104.1 | - | 3, 4 | 378 | |||
AJ002105.1 | - | 3, 4 | 378 | |||
AM409094.1 | - | 3, 4 | 379 | |||
AY366243.1 | - | 3, 4 | 388 | |||
AY366244.1 | - | 3, 4 | 377 | |||
BC108917.1 | - | 3, 4 | 380 | 40017299 | ||
BC119631.1 | - | 3, 4 | 397 | 40115568 | ||
BC119632.1 | - | 3, 4 | 397 | 40115570 | ||
BC125146.1 | - | 2, 3 | 362 | |||
BC126856.1 | - | 3, 4 | 380 | 40017301 | ||
FR750994.1 | - | 3, 4 | 413 | |||
L41347.1 | - | 3, 4 | 397 | |||
U24079.1 | - | 3, 4 | 376 | |||
X89893.1 | - | 3, 4 | 378 |
Gene Symbol: KIR2DS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014513.2 | NP_055328.2 | 3, 4 | 412 | ||
GenBank mRNA | AF208054.1 | - | 3, 4 | 412 | ||
AF272389.1 | - | 3, 4 | 362 | |||
AY366247.1 | - | 3, 4 | 436 | |||
AY366248.1 | - | 3, 5 | 436 | |||
L76672.1 | - | 3, 4 | 389 |
Gene Symbol: KIR3DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242867.1 | NP_001229796.1 | 4, 5 | 696 | ||
NM_006737.3 | NP_006728.2 | 4, 5 | 696 | |||
XM_011526940.2 | XP_011525242.1 | 4, 5 | 820 | |||
XM_017026784.1 | XP_016882273.1 | 4, 5 | 820 | |||
GenBank mRNA | AF208683.1 | - | 2, 3 | 600 | ||
AF208684.1 | - | 2, 3 | 600 | |||
AF208685.1 | - | 2, 3 | 600 | |||
AF208686.1 | - | 2, 3 | 600 | |||
AF208687.1 | - | 2, 3 | 600 | |||
AF262965.1 | - | 2, 3 | 582 | |||
AF262966.1 | - | 2, 3 | 582 | |||
AF262967.1 | - | 4, 5 | 696 | |||
AF263617.1 | - | 4, 5 | 696 | |||
AJ276125.1 | - | 4, 5 | 664 | |||
AW951147.1 | - | 1, 2 | 321 | |||
AY059418.1 | - | 2, 3 | 594 | |||
AY059419.1 | - | 2, 3 | 594 | |||
AY059420.1 | - | 2, 3 | 594 | |||
AY366256.2 | - | 4, 5 | 675 | |||
AY789067.1 | - | 4, 5 | 663 | |||
AY789068.1 | - | 4, 5 | 663 | |||
AY789069.1 | - | 3, 4 | 633 | |||
AY789070.1 | - | 4, 5 | 663 | |||
AY789071.1 | - | 3, 4 | 633 | |||
BC105678.1 | - | 4, 5 | 688 | 40004817 | ||
BC107951.1 | - | 4, 5 | 688 | 40004820 | ||
DQ256504.1 | - | 2, 3 | 595 | |||
EF582384.1 | - | 4, 5 | 663 | |||
L41270.1 | - | 4, 5 | 682 | |||
L76665.1 | - | 4, 5 | 674 | |||
L76666.1 | - | 4, 5 | 674 | |||
U30272.1 | - | 4, 5 | 665 | |||
X93595.1 | - | 4, 5 | 693 | |||
X93596.1 | - | 4, 5 | 663 | |||
X94373.1 | - | 4, 5 | 664 | |||
X94374.1 | - | 4, 5 | 663 |
Gene Symbol: KIR3DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083539.2 | NP_001077008.1 | 4, 5 | 700 | ||
NM_001282170.1 | NP_001269099.1 | 3, 4 | 415 | |||
NM_001282171.1 | NP_001269100.1 | 2, 3 | 379 | |||
GenBank mRNA | AF022044.1 | - | 4, 5 | 676 | ||
AJ417558.3 | - | 2, 3 | 612 | |||
AK301140.1 | - | 4, 5 | 700 | |||
BC098107.1 | - | 2, 3 | 353 | 40026977 | ||
BC098288.1 | - | 4, 5 | 674 | 40026980 | ||
BC105679.1 | - | 4, 5 | 674 | 40004313 | ||
EF370406.1 | - | 3, 4 | 623 | |||
EF582383.1 | - | 4, 5 | 663 | |||
L76661.1 | - | 4, 5 | 674 | |||
U73396.1 | - | 4, 5 | 663 | |||
X97232.1 | - | 3, 4 | 378 | |||
X97233.1 | - | 4, 5 | 663 |