Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR2DS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012313.1 | NP_036445.1 | 3 | 315 | ||
GenBank mRNA | AF022047.1 | - | 3 | 308 | ||
AM851002.1 | - | 3 | 340 | |||
AY789063.2 | - | 2 | 288 | |||
L76670.1 | - | 3 | 315 | |||
X97231.1 | - | 3 | 363 |
Gene Symbol: 1060P11.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ630586.1 | - | 4 | 590 |
Gene Symbol: KIR2DL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF135556.1 | - | 2 | 241 |
Gene Symbol: KIR2DS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281971.1 | NP_001268900.1 | 3 | 393 | ||
NM_001281972.1 | NP_001268901.1 | 3 | 393 | |||
NM_012314.5 | NP_036446.3 | 3 | 393 | |||
GenBank mRNA | AF002255.1 | - | 3 | 350 | ||
AF135564.1 | - | 2 | 145 | |||
AJ417554.1 | - | 2 | 284 | |||
AJ417555.1 | - | 2 | 284 | |||
AJ620616.2 | - | 2 | 277 | |||
AY102623.1 | - | 3 | 304 | |||
AY366245.1 | - | 3 | 338 | |||
AY388472.1 | - | 3 | 342 | |||
BC096694.1 | - | 3 | 315 | 40026979 | ||
BC101977.1 | - | 3 | 315 | 40004312 | ||
BC103693.1 | - | 3 | 315 | 40004314 | ||
BC108918.1 | - | 3 | 306 | 40017300 | ||
BC125147.1 | - | 2 | 288 | |||
BC127084.1 | - | 3 | 321 | 40120468 | ||
BC144566.1 | - | 3 | 393 | 9053094 | ||
BC171788.1 | - | 3 | 393 | 9054442 | ||
L76671.1 | - | 3 | 315 | |||
U24077.1 | - | 3 | 295 | |||
U96188.1 | - | 1 | 241 | |||
X94609.1 | - | 3 | 359 |