Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022075.4 | NP_071358.1 | 8 | 1015 | ||
NM_181746.3 | NP_859530.1 | 8 | 1055 | |||
XM_011509451.2 | XP_011507753.1 | 8 | 718 | |||
XM_011509452.2 | XP_011507754.1 | 8 | 766 | |||
GenBank mRNA | AF177338.2 | - | 8 | 744 | ||
AF189062.4 | - | 3 | 228 | |||
AK001105.1 | - | 7 | 684 | |||
AK290989.1 | - | 8 | 691 | |||
AY091458.1 | - | 8 | 837 | |||
BC001357.2 | - | 8 | 710 | 3051086 | ||
BC010032.2 | - | 8 | 674 | 3506408 | ||
BU543940.1 | - | 8 | 806 |