Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286067.1 | NP_001272996.1 | 4 | 468 | ||
NM_001286068.1 | NP_001272997.1 | 3 | 331 | |||
NM_001286069.1 | NP_001272998.1 | 3 | 336 | |||
NM_001286070.1 | NP_001272999.1 | 2 | 184 | |||
NM_001286071.1 | NP_001273000.1 | 3 | 336 | |||
NM_014039.3 | NP_054758.2 | 3 | 336 | |||
XM_006718824.1 | XP_006718887.1 | 4 | 377 | |||
XM_011542781.2 | XP_011541083.1 | 3 | 650 | |||
XM_011542782.2 | XP_011541084.1 | 3 | 741 | |||
XM_017017613.1 | XP_016873102.1 | 4 | 3792 | |||
XM_017017614.1 | XP_016873103.1 | 4 | 619 | |||
XM_017017615.1 | XP_016873104.1 | 4 | 1072 | |||
XM_017017616.1 | XP_016873105.1 | 3 | 328 | |||
XM_017017617.1 | XP_016873106.1 | 3 | 751 | |||
XM_017017618.1 | XP_016873107.1 | 3 | 327 | |||
XM_017017619.1 | XP_016873108.1 | 2 | 182 | |||
GenBank mRNA | AF092133.1 | - | 3 | 231 | ||
AK225977.1 | - | 3 | 266 | |||
AK291833.1 | - | 3 | 242 | |||
AK292215.1 | - | 4 | 387 | |||
AL136605.1 | - | 3 | 262 | |||
AL708049.1 | - | 3 | 343 | |||
AY203960.1 | - | 2 | 211 | |||
BC007110.1 | - | 2 | 91 | |||
BC012298.1 | - | 3 | 303 | 3852558 | ||
BI597310.1 | - | 2 | 106 | |||
BM551310.1 | - | 3 | 288 | |||
CR533538.1 | - | 2 | 104 |