Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014049.4 | NP_054768.2 | 2, 3 | 453 | ||
NR_033426.1 | 2, 3 | 629 | ||||
XM_017006264.1 | XP_016861753.1 | 2, 3 | 617 | |||
GenBank mRNA | AB209427.1 | - | 2, 3 | 2146 | ||
AF327351.1 | - | 2, 3 | 283 | |||
AK024012.1 | - | 2, 3 | 297 | |||
AK130311.1 | - | 2, 3 | 324 | |||
AK294597.1 | - | 2, 3 | 284 | |||
AK297952.1 | - | 2, 3 | 497 | |||
BC007970.2 | - | 2, 3 | 330 | 4304209 | ||
BC013354.2 | - | 2, 3 | 307 | 3935925 |