Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286067.1 | NP_001272996.1 | 4 | 501 | ||
NM_001286068.1 | NP_001272997.1 | 3 | 364 | |||
NM_001286069.1 | NP_001272998.1 | 3 | 369 | |||
NM_001286070.1 | NP_001272999.1 | 2 | 217 | |||
NM_001286071.1 | NP_001273000.1 | 3 | 369 | |||
NM_014039.3 | NP_054758.2 | 3 | 369 | |||
XM_006718824.1 | XP_006718887.1 | 4 | 410 | |||
XM_011542781.2 | XP_011541083.1 | 3 | 683 | |||
XM_011542782.2 | XP_011541084.1 | 3 | 774 | |||
XM_017017613.1 | XP_016873102.1 | 4 | 3825 | |||
XM_017017614.1 | XP_016873103.1 | 4 | 652 | |||
XM_017017615.1 | XP_016873104.1 | 4 | 1105 | |||
XM_017017616.1 | XP_016873105.1 | 3 | 361 | |||
XM_017017617.1 | XP_016873106.1 | 3 | 784 | |||
XM_017017618.1 | XP_016873107.1 | 3 | 360 | |||
XM_017017619.1 | XP_016873108.1 | 2 | 215 | |||
GenBank mRNA | AK225977.1 | - | 3 | 299 | ||
AK291833.1 | - | 3 | 275 | |||
AK292215.1 | - | 4 | 420 | |||
AL136605.1 | - | 3 | 295 | |||
AL708049.1 | - | 3 | 376 | |||
AY203960.1 | - | 2 | 244 | |||
BC007110.1 | - | 2 | 124 | |||
BC012298.1 | - | 3 | 336 | 3852558 | ||
BM551310.1 | - | 3 | 321 | |||
CR533538.1 | - | 2 | 137 |