Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020666.2 | NP_065717.1 | 2 | 249 | ||
GenBank mRNA | AF294429.1 | - | 1 | 141 | ||
AK294921.1 | - | 2 | 248 | |||
AK304614.1 | - | 2 | 251 | |||
AK308578.1 | - | 2 | 282 | |||
AK308674.1 | - | 2 | 308 | |||
AK308879.1 | - | 2 | 278 | |||
AK309215.1 | - | 2 | 278 | |||
AK310427.1 | - | 2 | 231 | |||
AK311485.1 | - | 2 | 251 | |||
BC015942.1 | - | 2 | 249 | |||
BC034633.1 | - | 2 | 203 | |||
BC065732.1 | - | 2 | 230 | |||
BC136261.1 | - | 2 | 197 | 9020248 | ||
BC143547.1 | - | 2 | 197 | 9052058 | ||
BC151233.1 | - | 2 | 284 | |||
CR749504.1 | - | 2 | 225 |