Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHEK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114121.2 | NP_001107593.1 | 11 | 2018 | ||
NM_001114122.2 | NP_001107594.1 | 11 | 2018 | |||
NM_001244846.1 | NP_001231775.1 | 11 | 2018 | |||
NM_001274.5 | NP_001265.2 | 11 | 1361 | |||
NR_045204.1 | 10 | 1794 | ||||
NR_045205.1 | 11 | 1555 | ||||
XM_011542560.2 | XP_011540862.1 | 10 | 2020 | |||
XM_011542561.2 | XP_011540863.1 | 10 | 2020 | |||
XM_011542562.2 | XP_011540864.1 | 10 | 1270 | |||
XM_011542563.2 | XP_011540865.1 | 10 | 1335 | |||
XM_017017146.1 | XP_016872635.1 | 11 | 1205 | |||
GenBank mRNA | AB451222.1 | - | 10 | 1121 | ||
AB451345.1 | - | 10 | 1121 | |||
AF016582.1 | - | 10 | 1155 | |||
AF032874.1 | - | 10 | 1121 | |||
AK292549.1 | - | 11 | 1555 | |||
AK293143.1 | - | 11 | 1183 | |||
AK299783.1 | - | 10 | 955 | |||
AK300082.1 | - | 10 | 1204 | |||
BC004202.2 | - | 11 | 1180 | 3530606 | ||
BC017575.1 | - | 11 | 1279 | 4091997 | ||
JF289264.1 | - | 10 | 996 |