Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166691.1 | NP_001160163.1 | 18 | 2202 | ||
NM_003318.4 | NP_003309.2 | 18 | 2193 | |||
XM_011536099.2 | XP_011534401.1 | 18 | 2167 | |||
XM_011536100.2 | XP_011534402.1 | 18 | 2190 | |||
XM_017011242.1 | XP_016866731.1 | 19 | 2722 | |||
GenBank mRNA | AK292460.1 | - | 18 | 2165 | ||
AK315696.1 | - | 18 | 2110 | |||
AX775733.1 | - | 18 | 2165 | |||
BC000633.2 | - | 18 | 2156 | 3343925 | ||
BC032858.2 | - | 18 | 2158 | 5270012 | ||
X70500.1 | - | 5 | 615 |