Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282161.1 | NP_001269090.1 | 6 | 765 | ||
NM_003336.3 | NP_003327.2 | 6 | 772 | |||
NM_181762.2 | NP_861427.1 | 5 | 682 | |||
GenBank mRNA | AK223045.1 | - | 6 | 674 | ||
AK223064.1 | - | 6 | 671 | |||
AK297696.1 | - | 5 | 584 | |||
AK301675.1 | - | 5 | 683 | |||
BC010175.2 | - | 6 | 665 | 3051041 | ||
BC042021.1 | - | 3 | 1549 | |||
BI093289.1 | - | 5 | 472 | |||
M74524.1 | - | 6 | 647 |