Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003482.3 | NP_003473.3 | 10 | 1527 | ||
XM_005269162.4 | XP_005269219.1 | 11 | 2806 | |||
XM_006719614.3 | XP_006719677.1 | 11 | 2806 | |||
XM_006719616.3 | XP_006719679.1 | 11 | 2806 | |||
XM_011538770.2 | XP_011537072.1 | 11 | 2809 | |||
XM_011538771.2 | XP_011537073.1 | 11 | 2809 | |||
XM_011538772.2 | XP_011537074.1 | 11 | 2809 | |||
XM_011538773.2 | XP_011537075.1 | 11 | 2809 | |||
XM_011538774.2 | XP_011537076.1 | 11 | 2810 | |||
XM_011538776.2 | XP_011537078.1 | 11 | 2809 | |||
GenBank mRNA | AF010403.1 | - | 10 | 1527 |