Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004964.2 | NP_004955.2 | 3, 4 | 338 | ||
XM_011541309.2 | XP_011539611.1 | 3, 4 | 342 | |||
GenBank mRNA | AB451297.1 | - | 3, 4 | 275 | ||
AB451430.1 | - | 3, 4 | 275 | |||
AK225555.1 | - | 3, 4 | 343 | |||
AK296308.1 | - | 2, 3 | 228 | |||
AK299241.1 | - | 3, 4 | 272 | |||
AK299792.1 | - | 3, 4 | 298 | |||
AK313061.1 | - | 3, 4 | 341 | |||
AY627042.1 | - | 3, 4 | 282 | |||
BC000301.2 | - | 3, 4 | 309 | 2820260 | ||
CR541717.1 | - | 3, 4 | 275 | |||
D50405.1 | - | 3, 4 | 338 | |||
U50079.1 | - | 3, 4 | 385 |