Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007257.2 | NP_001007258.1 | 3 | 2035 | ||
NM_012231.4 | NP_036363.2 | 8 | 3295 | |||
NM_015866.4 | NP_056950.2 | 8 | 3295 | |||
XM_017002253.1 | XP_016857742.1 | 7 | 2517 | |||
XM_017002254.1 | XP_016857743.1 | 7 | 2517 | |||
XM_017002255.1 | XP_016857744.1 | 7 | 2527 | |||
XM_017002256.1 | XP_016857745.1 | 8 | 2596 | |||
XM_017002257.1 | XP_016857746.1 | 7 | 2525 | |||
XM_017002258.1 | XP_016857747.1 | 5 | 2227 | |||
XM_017002259.1 | XP_016857748.1 | 4 | 2056 | |||
XM_017002260.1 | XP_016857749.1 | 3 | 2422 | |||
XM_017002261.1 | XP_016857750.1 | 3 | 2137 | |||
XM_017002262.1 | XP_016857751.1 | 4 | 2056 | |||
XM_017002263.1 | XP_016857752.1 | 3 | 1950 | |||
GenBank mRNA | AB208877.1 | - | 3 | 2014 | ||
AK293725.1 | - | 4 | 1871 | |||
AK300441.1 | - | 4 | 1743 | |||
AK308135.1 | - | 4 | 1868 | |||
BX647310.1 | - | 4 | 2058 | |||
D45132.1 | - | 3 | 2016 | |||
U17838.1 | - | 8 | 3297 |