Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMYD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020197.2 | NP_064582.2 | 6, 7 | 630 | ||
GenBank mRNA | AF226053.1 | - | 6, 7 | 619 | ||
AK098683.1 | - | 2, 3 | 114 | |||
AK313868.1 | - | 6, 7 | 600 | |||
BC028104.1 | - | 5, 6 | 551 | |||
BC098133.1 | - | 6, 7 | 619 | 40005281 | ||
BC098276.1 | - | 6, 7 | 619 | 40005283 | ||
BC098305.1 | - | 6, 7 | 619 | 40005284 | ||
BC098335.1 | - | 6, 7 | 619 | 40005280 |