Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUAK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030952.2 | NP_112214.2 | 7 | 3428 | ||
GenBank mRNA | AL136891.1 | - | 7 | 3392 | ||
BC017306.2 | - | 7 | 3364 | 5088037 | ||
CR749209.1 | - | 3 | 6857 |