Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL12RB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290023.1 | NP_001276952.1 | 9 | 1175 | ||
NM_001290024.1 | NP_001276953.1 | 10 | 1166 | |||
NM_005535.2 | NP_005526.1 | 9 | 1175 | |||
NM_153701.2 | NP_714912.1 | 9 | 1175 | |||
XM_006722741.3 | XP_006722804.2 | 10 | 1368 | |||
XM_011527966.2 | XP_011526268.1 | 10 | 1410 | |||
XM_011527967.2 | XP_011526269.1 | 10 | 1396 | |||
XM_011527968.2 | XP_011526270.1 | 10 | 1383 | |||
XM_011527969.2 | XP_011526271.1 | 10 | 1369 | |||
XM_011527970.2 | XP_011526272.1 | 10 | 1409 | |||
XM_011527971.2 | XP_011526273.1 | 10 | 1407 | |||
XM_011527972.2 | XP_011526274.1 | 10 | 1407 | |||
XM_011527973.2 | XP_011526275.1 | 9 | 1201 | |||
XM_011527974.2 | XP_011526276.1 | 9 | 1189 | |||
XM_011527975.2 | XP_011526277.1 | 10 | 1366 | |||
XM_011527976.2 | XP_011526278.1 | 10 | 1407 | |||
XM_011527977.2 | XP_011526279.1 | 10 | 1359 | |||
XM_017026762.1 | XP_016882251.1 | 5 | 547 | |||
GenBank mRNA | AK225919.1 | - | 9 | 1106 | ||
AK290423.1 | - | 9 | 1103 | |||
BC029121.1 | - | 9 | 1064 | |||
BC137404.1 | - | 9 | 1020 | 9021402 | ||
BC137406.1 | - | 9 | 1020 | 9021404 | ||
BX647221.1 | - | 10 | 1166 | |||
U03187.1 | - | 9 | 1059 |