Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282454.1 | NP_001269383.1 | 9 | 1053 | ||
NM_014426.3 | NP_055241.1 | 9 | 1181 | |||
NM_152227.2 | NP_689413.1 | 10 | 1136 | |||
GenBank mRNA | AF121855.1 | - | 10 | 992 | ||
AK001793.1 | - | 9 | 898 | |||
AK026227.1 | - | 2 | 93 | |||
AK054634.1 | - | 10 | 1924 | |||
AK129505.1 | - | 9 | 898 | |||
AK223308.1 | - | 9 | 866 | |||
AK296947.1 | - | 10 | 1005 | |||
AK308017.1 | - | 10 | 1137 | |||
BC062638.1 | - | 9 | 838 | |||
BC093623.1 | - | 9 | 1107 | 7939468 | ||
BC093980.1 | - | 9 | 1107 | 7939825 | ||
BC143274.1 | - | 9 | 1107 | 9051776 |