Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NIPA2 SIRNA Assays ›
Gene Symbol:
NIPA2
Gene Name:
non imprinted in Prader-Willi/Angelman syndrome 2
Gene Aliases:
-
Chromosome Location:
Chr.15: 22838641 - 22868384 on Build GRCh38
UniGene ID:
Hs.591003
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
214841
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NIPA2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001008860.2
NP_001008860.1
7
2324
NM_001008892.2
NP_001008892.1
6
2188
NM_001008894.2
NP_001008894.1
5
2131
NM_001184888.1
NP_001171817.1
6
2279
NM_001184889.1
NP_001171818.1
10
2671
NM_030922.6
NP_112184.4
8
2446
XM_005272546.3
XP_005272603.1
8
2679
XM_005272547.4
XP_005272604.1
8
2463
XM_005272548.3
XP_005272605.1
7
2293
XM_005272550.3
XP_005272607.1
7
2131
XM_005272552.3
XP_005272609.1
6
2393
XM_005272553.4
XP_005272610.1
6
2197
XM_006720364.2
XP_006720427.1
9
2573
XM_006720365.2
XP_006720428.1
8
2418
XM_006720366.3
XP_006720429.1
7
2284
XM_006720367.1
XP_006720430.1
7
2622
XM_011543877.2
XP_011542179.1
9
2806
XM_011543878.2
XP_011542180.1
9
2973
XM_011543879.2
XP_011542181.1
9
2544
XM_011543880.2
XP_011542182.1
7
2557
XM_017022645.1
XP_016878134.1
9
2867
XM_017022646.1
XP_016878135.1
9
2634
XM_017022647.1
XP_016878136.1
9
2582
XM_017022648.1
XP_016878137.1
9
2643
XM_017022649.1
XP_016878138.1
9
2777
XM_017022650.1
XP_016878139.1
9
2740
XM_017022651.1
XP_016878140.1
10
3161
XM_017022652.1
XP_016878141.1
9
2553
XM_017022653.1
XP_016878142.1
10
2867
XM_017022654.1
XP_016878143.1
8
2391
XM_017022655.1
XP_016878144.1
7
2389
XM_017022656.1
XP_016878145.1
7
2399
XM_017022657.1
XP_016878146.1
8
2516
XM_017022658.1
XP_016878147.1
6
2237
XM_017022659.1
XP_016878148.1
8
2487
XM_017022660.1
XP_016878149.1
6
2500
XM_017022661.1
XP_016878150.1
6
2267
XM_017022662.1
XP_016878151.1
5
2336
XM_017022663.1
XP_016878152.1
5
2141
GenBank mRNA
AK096305.1
-
8
2202
AY732242.1
-
10
2592
BC000957.3
-
3
1618
BC011775.2
-
6
2100
3640970
U90904.1
-
1
1280
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
magnesium ion transport
magnesium ion transmembrane transport
magnesium ion transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-lhtlm:80/10.239.167.245:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0