Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BIN1 SIRNA Assays ›
Gene Symbol:
BIN1
Gene Name:
bridging integrator 1
Gene Aliases:
AMPH2, AMPHL, SH3P9
Chromosome Location:
Chr.2: 127048023 - 127107400 on Build GRCh38
UniGene ID:
Hs.193163
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
214862
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BIN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320632.1
NP_001307561.1
6
890
NM_001320633.1
NP_001307562.1
6
890
NM_001320634.1
NP_001307563.1
6
656
NM_001320640.1
NP_001307569.1
6
890
NM_001320641.1
NP_001307570.1
6
890
NM_001320642.1
NP_001307571.1
6
1384
NM_004305.3
NP_004296.1
6
890
NM_139343.2
NP_647593.1
6
890
NM_139344.2
NP_647594.1
6
890
NM_139345.2
NP_647595.1
6
890
NM_139346.2
NP_647596.1
6
890
NM_139347.2
NP_647597.1
6
890
NM_139348.2
NP_647598.1
6
890
NM_139349.2
NP_647599.1
6
890
NM_139350.2
NP_647600.1
6
890
NM_139351.2
NP_647601.1
6
890
XM_005263642.3
XP_005263699.1
6
963
XM_005263643.3
XP_005263700.1
6
963
XM_005263645.2
XP_005263702.1
6
447
XM_006712425.3
XP_006712488.1
6
963
XM_006712426.3
XP_006712489.1
6
963
XM_006712427.2
XP_006712490.1
6
447
XM_006712428.3
XP_006712491.1
6
963
XM_006712429.3
XP_006712492.1
6
963
XM_006712431.3
XP_006712494.1
6
963
XM_006712432.3
XP_006712495.1
6
963
XM_006712433.3
XP_006712496.1
6
963
XM_006712434.3
XP_006712497.1
6
963
XM_011510975.2
XP_011509277.1
6
1596
XM_017003819.1
XP_016859308.1
6
777
XM_017003820.1
XP_016859309.1
6
963
XM_017003821.1
XP_016859310.1
6
963
XM_017003822.1
XP_016859311.1
6
963
XM_017003823.1
XP_016859312.1
6
447
XM_017003824.1
XP_016859313.1
6
447
XM_017003825.1
XP_016859314.1
6
777
XM_017003826.1
XP_016859315.1
6
447
XM_017003827.1
XP_016859316.1
6
963
XM_017003828.1
XP_016859317.1
6
963
GenBank mRNA
AF001383.1
-
6
677
AF004015.1
-
6
506
AF043898.1
-
6
506
AF043899.1
-
6
506
AF043900.1
-
6
506
AF043901.1
-
6
506
AF068914.1
-
6
566
AF068915.1
-
6
566
AF068916.1
-
6
566
AF068917.1
-
6
566
AF068918.1
-
6
566
AF070576.1
-
2
112
AK295300.1
-
6
608
AK301153.1
-
6
656
AL537027.3
-
6
644
AL713697.1
-
6
851
AW411140.1
-
2
161
BC004101.2
-
6
566
3688734
BT006865.1
-
6
506
U68485.1
-
6
552
U87558.1
-
6
527
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_053959, XM_006254505, XM_008772013, XM_008772014, XM_006254497, and others.This siRNA also targets the following transcripts in Mouse: NM_001083334, NM_009668, BU707946, U60884, BC065160.
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
nucleus organization
cell proliferation
viral process
regulation of endocytosis
muscle cell differentiation
positive regulation of apoptotic process
regulation of neuron differentiation
positive regulation of astrocyte differentiation
lipid tube assembly
regulation of cell cycle arrest
protein binding
identical protein binding
tau protein binding
actin filament binding
RNA polymerase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-c4z4p:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0