Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025577.2 | NP_001020748.2 | 1 | 3341 | ||
GenBank mRNA | AK033026.1 | - | 256 | |||
AK079582.1 | - | 1 | 1398 | |||
AK134055.1 | - | 1 | 1510 | |||
AK151743.1 | - | 1 | 1252 | |||
BC034073.1 | - | 1 | 1519 | |||
BC041683.1 | - | 1 | 1519 |