Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003088.3 | NP_003079.1 | 1 | 557 | ||
GenBank mRNA | AK095254.1 | - | 2 | 491 | ||
AK316607.1 | - | 1 | 552 | |||
BC000521.2 | - | 1 | 448 | 2961047 | ||
BC006304.2 | - | 1 | 539 | 4100996 | ||
BC007539.1 | - | 1 | 448 | |||
BC007643.1 | - | 1 | 181 | |||
BC007948.2 | - | 1 | 536 | 4299524 | ||
BC007988.2 | - | 1 | 448 | 2961047 | ||
BI546742.1 | - | 1 | 556 | |||
BT006636.1 | - | 1 | 425 | |||
U03057.1 | - | 1 | 536 | |||
U09873.1 | - | 1 | 470 |