Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005967.3 | NP_005958.1 | 2 | 757 | ||
XM_005268894.2 | XP_005268951.1 | 2 | 757 | |||
GenBank mRNA | AK314229.1 | - | 2 | 537 | ||
BC007756.2 | - | 2 | 491 | |||
BC065931.1 | - | 2 | 544 | 6157017 | ||
U48361.1 | - | 2 | 536 | |||
X70991.1 | - | 1 | 232 |