Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003592.2 | NP_003583.2 | 17 | 2235 | ||
XM_005250060.4 | XP_005250117.1 | 17 | 2271 | |||
XM_011516629.2 | XP_011514931.1 | 17 | 2227 | |||
XM_011516630.2 | XP_011514932.1 | 17 | 2128 | |||
XM_011516631.2 | XP_011514933.1 | 17 | 2256 | |||
XM_011516632.2 | XP_011514934.1 | 17 | 2154 | |||
XM_017012723.1 | XP_016868212.1 | 16 | 1991 | |||
GenBank mRNA | AF062536.1 | - | 16 | 2009 | ||
AK096163.1 | - | 18 | 2123 | |||
AK096505.1 | - | 10 | 1892 | |||
AK296909.1 | - | 16 | 1960 | |||
AK315023.1 | - | 17 | 2411 | |||
BC125119.1 | - | 17 | 2148 | 40118923 | ||
BC125120.1 | - | 17 | 2148 | 40118927 | ||
BX537409.1 | - | 17 | 2165 | |||
U58087.1 | - | 17 | 1937 |