Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184728.2 | NP_001171657.1 | 7 | 803 | ||
NM_001184729.2 | NP_001171658.1 | 8 | 971 | |||
NM_001184730.2 | NP_001171659.1 | 6 | 665 | |||
NM_015937.5 | NP_057021.2 | 8 | 971 | |||
NR_047691.1 | 8 | 931 | ||||
NR_047692.1 | 7 | 874 | ||||
NR_047693.1 | 7 | 870 | ||||
NR_047694.1 | 7 | 793 | ||||
NR_047695.1 | 5 | 564 | ||||
XM_005260430.2 | XP_005260487.1 | 6 | 643 | |||
XM_005260432.2 | XP_005260489.1 | 7 | 765 | |||
GenBank mRNA | AB057724.1 | - | 8 | 881 | ||
AF058295.1 | - | 1, 2 | 180 | |||
AF132940.1 | - | 8 | 875 | |||
AK075469.1 | - | 8 | 895 | |||
AK225517.1 | - | 8 | 901 | |||
AK293294.1 | - | 7 | 794 | |||
AK293686.1 | - | 7 | 716 | |||
AK296063.1 | - | 7 | 800 | |||
AK296139.1 | - | 7 | 726 | |||
AK297830.1 | - | 3 | 450 | |||
AK299013.1 | - | 5 | 490 | |||
AK301815.1 | - | 7 | 768 | |||
AK302093.1 | - | 6 | 623 | |||
AK311100.1 | - | 7 | 753 | |||
AK311416.1 | - | 6 | 615 | |||
AK311492.1 | - | 8 | 866 | |||
AK311520.1 | - | 6 | 667 | |||
AK311522.1 | - | 7 | 782 | |||
AK316358.1 | - | 8 | 857 | |||
AL121742.1 | - | 8 | 874 | |||
AY358588.1 | - | 8 | 878 | |||
BC015022.1 | - | 8 | 900 | 3921680 | ||
BC110892.1 | - | 8 | 879 | |||
BC136827.1 | - | 8 | 909 | 9020818 | ||
BC136828.1 | - | 8 | 909 | 9020819 | ||
BX537612.1 | - | 6 | 2309 |