Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015362.4 | NP_056177.3 | 5 | 486 | ||
NM_203413.2 | NP_981958.1 | 5 | 486 | |||
NM_203414.2 | NP_981959.1 | 4 | 574 | |||
NM_203415.1 | NP_981960.1 | 5 | 379 | |||
XM_011523779.2 | XP_011522081.1 | 5 | 478 | |||
GenBank mRNA | AB013910.1 | - | 4 | 272 | ||
AF070658.1 | - | 4 | 247 | |||
AF087868.1 | - | 4 | 247 | |||
AF163262.1 | - | 4 | 205 | |||
AK225983.1 | - | 4 | 489 | |||
AK289940.1 | - | 4 | 489 | |||
BC002762.2 | - | 5 | 451 | 3629369 | ||
BE743557.1 | - | 5 | 379 | |||
BI916017.1 | - | 4 | 571 | |||
BQ648358.1 | - | 5 | 476 |