Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166269.1 | NP_001159741.1 | 10 | 1594 | ||
NM_001166270.1 | NP_001159742.1 | 9 | 1451 | |||
NM_017815.2 | NP_060285.2 | 10 | 1586 | |||
GenBank mRNA | AK000431.1 | - | 10 | 1555 | ||
AK026016.1 | - | 10 | 1540 | |||
AK057779.1 | - | 8 | 1381 | |||
BC001916.1 | - | 10 | 1583 | 2905474 | ||
BC002554.2 | - | 10 | 1517 | 3139198 | ||
BX247981.1 | - | 7 | 1392 | |||
BX247995.1 | - | 9 | 1403 | |||
BX248256.1 | - | 9 | 1418 |