Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320871.1 | NP_001307800.1 | 7 | 1972 | ||
NM_201520.2 | NP_958928.1 | 6 | 3113 | |||
GenBank mRNA | AK097536.1 | - | 6 | 2622 | ||
BC041597.1 | - | 7 | 1972 | |||
BC052233.1 | - | 7 | 1972 |