Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006996.1 | NP_001006997.1 | 5 | 770 | ||
GenBank mRNA | BC083919.1 | - | 5 | 770 | 7135005 |