Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007639.1 | NP_001007640.1 | 14 | 2116 | ||
GenBank mRNA | BC079194.1 | - | 14 | 2116 | 7131380 |