Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1359634 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007708.1 | NP_001007709.1 | 6 | 904 | ||
XM_006241992.2 | XP_006242054.1 | 7 | 751 | |||
XM_008765713.1 | XP_008763935.1 | 8 | 777 | |||
GenBank mRNA | BC079039.1 | - | 6 | 904 | 7114722 |