Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcakd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007724.1 | NP_001007725.1 | 3 | 418 | ||
XM_006247532.2 | XP_006247594.1 | 3 | 667 | |||
XM_008768293.1 | XP_008766515.1 | 3 | 465 | |||
XM_008768294.1 | XP_008766516.1 | 3 | 783 | |||
GenBank mRNA | BC079186.1 | - | 3 | 418 | 7131172 |